Learn More
Selleck Chemical LLC BLU 451-E1877-25MG
SDP
Supplier:  Selleck Chemical LLC E187725MG
Encompass_Preferred
BLU 451 is a central nervous system (CNS)-penetrant wild-type EGFR-sparing covalent inhibitor of epidermal growth factor receptor (EGFR) exon 20 insertion (Ex20ins) with an IC50 of 7-78 nM It also exhibits a potent anti-tumor efficacy
Catalog No. 50-313-5781