Learn More
TARGETMOL CHEMICALS INC BLU-782 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T81085MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Blu-782 (ALK2-IN-1) is a activin receptor-like kinase-2 (ALK2) inhibitor ( IC50 of <10 nM)purity: 98%
Catalog No. 50-257-9103