Learn More
Medchemexpress LLC Blu2864 | 2810747-89-6 | 99.5% | 452.43 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY15007650MG
Encompass_Preferred
BLU2864 is an orally active, highly selective, and ATP-competitive PRKACA inhibitor with an IC50 of 0.3 nM. This compound demonstrates anti-tumor activity and is suitable for research related to cancer and polycystic kidney disease.
  • Highly selective, ATP-competitive PRKACA inhibitor
  • Exhibits anti-tumor activity
  • Supports cancer research
  • Supports polycystic kidney disease research
  • Inhibits forskolin-induced in vitro cystogenesis
  • Inhibits renal PKA activity
  • Ameliorates polycystic kidney disease
  • Reduces tumor growth in vivo
Catalog No. 50-003-61268