Learn More
Sigma Aldrich Fine Chemicals Biosciences Blue Dextran Void volume marker for gel filtration columns. | 87915-38-6 | MFCD00130661 | 10G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences D575110G
Encompass
Blue Dextran Void volume marker for gel filtration columns. | 87915-38-6 | MFCD00130661 | 10G
Catalog No. 50-178-1177