Learn More
Chemglass Life Sciences Cap, GL-45, Blue, Open Top, (1-5/16" Dia Hole) Polypropylene
SDP
Supplier:  Chemglass Life Sciences CLS148121
Encompass
Open top polypropylene caps for use with port assemblies.Caps can be autoclaved in steam to 140 degreesC.
Catalog No. 50-121-5138