Learn More
Bellco Glass, Inc. Blue Tube Marker
SDP
Supplier:  Bellco Glass, Inc. 229406000
Encompass_Preferred
DescriptionFast drying, smudge resistant, fine point markerIdeal for permanently marking all plastic lab ware, especially centrifuge tubes; even works with frozen tubes! Pack Description5/BagCase Quantity5Primary ColorBlueAlcohol/Solvent ResistantNo
Catalog No. 50-233-0292