Learn More
Gold Biotechnology Inc BLUEstainTM 2 Protein ladder
SDP
Supplier:  Gold Biotechnology Inc P0083000
Encompass_Preferred
The BLUEstain 2 Protein ladder is a three-color protein standard with 13 prestained proteins covering a wide range molecular weights from 3 5 to 245 kDa Proteins are covalently coupled with a blue chromophore except for two reference bands (one green and one red band at 25 kDa and 75 kDa respectively) when separated on SDS-PAGE (Tris-glycine buffer)
Catalog No. 50-005-62101