Learn More
Scientific Commodities Inc BLUNT TIP PIPETTING NDL 3PK
Supplier:  Scientific Commodities Inc BB8294223
Encompass_Preferred
BLUNT TIP LABORATORY PIPETTING NEEDLES. Pack of 3Needle Gauge: 22Needle Dimensions: .016″ ID x .028″ ODLength: 4 Inch
Catalog No. NC1788117