Learn More
Cayman Chemical BM 567 (TXA2), 1MG
SDP
Supplier:  Cayman Chemical 101551
Encompass_Preferred
BM 567 Thromboxane A2 (TXA2) is a potent thrombogenic and vasoconstrictor eicosanoid, produced in large quantities by activated platelets. TXA2 has been implicated as a causal factor in the onset of stroke and myocardial infarction. BM 567 is a dual acting antithrombogenic agent, acting as an inhibitor of thromboxane A2 (TXA2) synthase and as an antagonist of the TP receptor, the G protein-coupled receptor mediatingTXA2 activity in platelets and vascular smooth muscle. BM 567antagonizes the vascular smooth muscle TP receptor with an IC50 value of1.1 nM. It inhibits platelet TX synthase with an IC50 value of 12 nM.
Catalog No. NC2223180