Learn More
Selleck Chemical LLC BMH-21 10mg 896705-16-1
SDP
Supplier:  Selleck Chemical LLC S771810MG
Encompass_Preferred
BMH-21 is a DNA intercalator, which binds ribosomal DNA and inhibits RNA polymerase I (Pol I) transcription. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5165