Learn More
TARGETMOL CHEMICALS INC BMH-21 50MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T176750MG
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BMH-21 a small molecule DNA intercalator binds ribosomal DNA and inhibits RNA polymerase I (Pol I) transcription and not affects phosphorylation of H2AX. purity: 99%
Catalog No. 50-258-1097