Learn More
Medchemexpress LLC BMH-21 | 896705-16-1 | 99.3% | 360.41 | C21H20N4O2 | 500MG
SDP
Supplier:  Medchemexpress LLC HY12484500MG
Encompass_Preferred
BMH-21 is a planar heterocyclic small-molecule DNA intercalator that selectively inhibits RNA polymerase I (Pol I) transcription and demonstrates anticancer activity. It binds GC-rich ribosomal DNA, induces nucleolar stress, and promotes degradation of the Pol I subunit RPA194, resulting in suppression of ribosomal RNA synthesis. Supplied as a research-grade powder for in vitro and preclinical studies.
  • DNA intercalator with preference for GC-rich sequences.
  • Selective inhibitor of RNA polymerase I transcription.
  • Induces nucleolar stress and promotes RPA194 degradation.
  • High purity research-grade material suitable for biochemical studies.
  • Supplied as a powder with recommended cold storage for stability.
  • Commonly used in cancer biology and transcription research.
Catalog No. 50-002-39259