Learn More
Apexbio Technology LLC BML-210(CAY10433), 537034-17-6, MFCD08062139, 10 mg
SDP
Supplier:  Apexbio Technology LLC B596810
Encompass_Preferred
MF: C20H25N3O2, MW: 339.43, Solubility: 10.5mg/mL in DMSO. BML-210 (CAY10433) is a novel HDAC inhibitor with IC50 value of 87 uM.
Catalog No. 50-115-2419