Learn More
Abcam BML-210, HDAC inhibitor, 1MG
Supplier:  Abcam AB1416401MG
Encompass_Preferred
MW 339.4 Da, Purity >98%. Histone deacetylase (HDAC) inhibitor (IC₅₀ = 87 μM). Induces changes in gene expression profile. Antiproliferative activity in vitro.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6485