Learn More
Medchemexpress LLC BML-260 | 101439-76-3 | 99.7% | 341.40 g/mol | C17H11NO3S2 | 5 MG

Supplier: Medchemexpress LLC HY1330955MG
BML-260 is a small-molecule inhibitor of the dual-specificity phosphatase DUSP22 (JSP-1) used in biochemical and cellular research. It modulates JNK-related signaling and has been reported to activate UCP1 and influence CREB, STAT3, and PPAR pathways in adipocytes.
- Potent DUSP22 (JSP-1) inhibitor for biochemical assays.
- Useful for research on inflammation, proliferative disorders, and adipocyte thermogenesis.
- High purity (~99.7%) suitable for research applications.
- Molecular weight 341.40 g/mol for analytical reference.
- Available in small research quantities, for example 5 mg.
- CAS number 101439-76-3 for unambiguous identification.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.