Learn More
Enzo Life Sciences BML-260 (10mg). CAS: 101439-76-3
SDP
Supplier:  Enzo Life Sciences BMLPR1190010
Encompass_Preferred
A potent inhibitor of the dual specificity phosphatase JSP-1 (IC50=18µM). Purity: ≥98% (TLC). Solubility: Soluble in DMSO (25mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-1514