Learn More
Abcam BML 284, Wnt signaling activator, 5MG
Supplier:  Abcam AB1452415MG
Encompass_Preferred
MW 350.4 Da, Purity >98%. Cell-permeable Wnt signaling activator. Induces β-catenin and TCF-dependent transcriptional activity (EC₅₀ = 0.7 μM). Activates Wnt without inhibiting GSK-3β.Shows developmental effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5365