Learn More
STEMCELL Technologies BMN 673, Size: 10 mg
SDP
Supplier:  STEMCELL Technologies 1001129
Encompass
Inhibit poly ADP ribose polymerase (PARP) with BMN 673 (PARP1 IC50 0.57 nM Shen et al.). BMN 673 binds to PARP in cancer cells with BRCA1 BRCA2 or PTEN mutations and prevents PARP-mediated repair of single-strand DNA breaks leading to DNA damage (Shen et al.).
Catalog No. 50-239-0175