Learn More
Enzo Life Sciences BMPO (high purity) (10mg)
SDP
Supplier:  Enzo Life Sciences ALX430141M010
Encompass_Preferred
Nitrone spin trap for the specific in vivo or in vitro detection of short-lived superoxide, hydroxyl and thiyl radicals. Forms distinguishable adducts which can be measured by EPR spectroscopy. Unlike with DMPO, the superoxide adduct does not decay into a hydroxyl adduct and it has a much longer half-life (t1/2=23min). Low paramagnetic impurities. No further purification required. Alternative name: 5-tert-Butoxycarbonyl-5-methyl-1-pyrroline-N-oxide. Purity: ≥99% (HPLC). Appearance: White to pinkish crystalline solid. Solubility: Soluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-9231