Learn More
Cayman Chemical BmrABsubtIlIs 168 10ug
SDP
Supplier:  Cayman Chemical 2473410 UG
Encompass_Preferred
Source: N-terminally histidine-tagged BmrA (B. subtilis strain 168) purified from E. coli • AAs: Full-length, wild-type sequence • MW = 64.5 kDa
Catalog No. 50-301-5842