Learn More
Selleck Chemical LLC BMS-1166 S8859-25MG
SDP
Supplier:  Selleck Chemical LLC S885925MG
Encompass_Preferred
BMS-1166 is a novel and potent small molecule inhibitor of the PD-1/PD-L1 interaction with an IC50 of 1 4 nM
Catalog No. 50-000-25386