Learn More
Medchemexpress LLC BMS-1166 | 1818314-88-3 | 99.5% | 641.11 | C36H33ClN2O7 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY102011100MG
Encompass_Preferred
BMS-1166 is a small-molecule inhibitor of the PD-1/PD-L1 protein-protein interaction used in research to block immune checkpoint signaling. It is supplied as a white to off-white solid with high purity and is intended for in vitro and cellular studies of PD-L1 biology and immune modulation.
  • Potent low-nanomolar PD-1/PD-L1 inhibitory activity (IC50 ≈ 1.4 nM).
  • High purity, about 99.5% by HPLC.
  • Chemical formula C36H33ClN2O7, molecular weight 641.11 g·mol-1.
  • White to off-white solid; soluble in DMSO.
  • Suitable for biochemical and cell-based assays targeting PD-L1.
Catalog No. 50-002-47393