Learn More
Medchemexpress LLC BMS-1166 | 1818314-88-3 | 99.5% | 641.11 g·mol⁻¹ | C36H33ClN2O7 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10201125MG
Encompass_Preferred
BMS-1166 is a small-molecule inhibitor of the PD-1/PD-L1 immune checkpoint that induces PD-L1 dimerization and blocks PD-1/PD-L1 interaction. It is supplied for research use and is provided in milligram-scale quantities suitable for biochemical and cellular assays.
  • Potent PD-1/PD-L1 inhibitor with reported IC50 of 1.4 nM.
  • High purity suitable for biochemical and cellular assays.
  • Available in small milligram pack sizes for flexible dosing.
  • Soluble in DMSO for assay preparation.
  • Stable as a powder when stored at -20°C per manufacturer guidance.
Catalog No. 50-002-47579