Learn More
Medchemexpress LLC BMS-1166 | 1818314-88-3 | MFCD31746874 | ≥98.0% | 641.11 g·mol⁻¹ | C36H33ClN2O7 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1020111MG
Encompass_Preferred
BMS-1166 is a small-molecule inhibitor of the PD-1/PD-L1 protein-protein interaction used in preclinical immuno-oncology research. It binds PD-L1, induces PD-L1 dimerization, and blocks PD-1 binding, restoring T-cell activation in cellular assays. Reported identifiers include CAS 1818314-88-3, formula C36H33ClN2O7, and molecular weight 641.11 g·mol⁻¹.
  • Potent PD-1/PD-L1 interaction inhibition with IC50 ≈ 1.4 nM.
  • Demonstrated cellular activity (EC50 ≈ 276 nM in a Jurkat/CHO reporter assay).
  • High purity suitable for research use (≥98%).
  • Solid powder formulation, stored at -20°C for long-term stability.
  • Commonly used for in vitro immune checkpoint blockade and preclinical assays.
Catalog No. 50-002-47533