Learn More
Medchemexpress LLC BMS-1166-N-piperidine-COOH | 2447066-00-2 | 97.2% | C37H35ClN2O6 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13118710MG
Encompass_Preferred
BMS-1166-N-piperidine-COOH is a research-grade small-molecule fragment that functions as a PD-1/PD-L1 interaction inhibitor ligand and a synthetic building block for assembling PROTAC degraders and related modulators. Supplied as a characterized solid with reported high purity and defined molecular composition, it is intended for laboratory research use only.
  • High purity (97.2%) suitable for research synthesis and screening.
  • Defined molecular formula (C37H35ClN2O6) and molecular weight (~639.14 g·mol⁻¹).
  • Designed for use as a building block in PROTAC and PD-1/PD-L1 modulator development.
  • Available in small-scale package sizes for early-stage research.
  • Supplied for research use only; not for clinical or human use.
Catalog No. 50-002-61550