Learn More
Cayman Chemical BMS 1166hydrochlorIde 25mg
SDP
Supplier:  Cayman Chemical 2777425 MG
Encompass_Preferred
An inhibitor of the interaction between PD-1 and PDL1 (IC50 = 1.4 nM in a HTRF assay); increases the activation of Jurkat cells expressing PD-1 in co-culture with CHO cells expressing PD-L1 (EC50 = 276 nM in a reporter assay)
Catalog No. 50-294-0825