Learn More
Medchemexpress LLC BMS 182874 | 153042-42-3 | 345.42 | C17H19N3O3S | 1 ML
SDP
Supplier:  Medchemexpress LLC HY11849710MM 1ML
Encompass_Preferred
BMS 182874 is an orally active, highly selective endothelin A (ETA) receptor antagonist used in cardiovascular and GPCR research. Supplied as a 10 mM solution (1 mL) for laboratory research use only. Reported potency: ETA IC50 0.15 μM and Ki 0.055 μM. Molecular formula C17H19N3O3S; molecular weight 345.42. Store protected from light; in solvent store -80°C for up to 6 months.
  • Provided as 10 mM solution in DMSO, 1 mL.
  • Selective ETA receptor antagonist with ETA IC50 0.15 μM and Ki 0.055 μM.
  • Validated in vsm-A10 cell assays for binding and Ca2+ antagonism.
  • High solubility in DMSO; ultrasonic treatment may be required.
  • Intended for laboratory research use only; not for human or veterinary use.
Catalog No. 50-002-26164