Learn More
Medchemexpress LLC BMS 182874 | 153042-42-3 | 99.9% | 345.42 g·mol⁻¹ | C17H19N3O3S | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11849710MG
Encompass_Preferred
BMS-182874 is a selective endothelin ETA receptor antagonist used as a research compound in cardiovascular and pulmonary pharmacology. It is supplied as a high-purity solid suitable for in vitro assays and preclinical pharmacology studies.
  • Selective ETA receptor antagonist with reported IC50 0.150 μM and Ki 0.055 μM.
  • High reported purity (~99.9%).
  • Supplied as a solid in a 10 mg quantity.
  • Intended for use in cardiovascular and hypertension research models.
  • For research use only; not for human or veterinary use.
Catalog No. 50-002-27454