Learn More
Medchemexpress LLC BMS-192364 25mg | 202822-21-7 | 25 MG

Supplier: Medchemexpress LLC HY11486525MG
BMS-192364 is a small-molecule regulator of G protein signaling (RGS) agonist for research use. It enhances GAP activity on Gq proteins, reduces urinary bladder contraction, and inhibits calcium flux. The compound is supplied as a white to off-white solid with high purity and recommended low-temperature storage.
- High purity: 99.01% as reported by supplier.
- Molecular weight: 355.70 g/mol.
- Solid form; white to off-white powder suitable for biochemical and cellular assays.
- Recommended storage: powder at -20°C for long-term stability; in solvent at -80°C for extended storage.
- Available in a 25 mg pack size for small-scale research applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.