Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bms-199264 (hydrochloride) | 186180-83-6 | 99.3% | 567.53 | C26H32Cl2N4O4S | 10 MG
SDP

Supplier:  Medchemexpress LLC HY11896010MG

Encompass_Preferred

This hydrochloride salt is a small-molecule inhibitor of mitochondrial F1F0 ATP synthase supplied as a white to off-white solid for laboratory research use. It inhibits ATP hydrolase activity and should be handled and stored according to supplier guidance.

  • Potent inhibitor of mitochondrial F1F0 ATP synthase activity.
  • High purity (≈99.3%) suitable for research applications.
  • Available in small-scale quantities for laboratory testing.
  • White to off-white solid; stable when stored sealed at 4°C.
  • Solvent stability: -80°C (6 months), -20°C (1 month) for prepared solutions.

Catalog No. 50-002-27396


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.