Learn More
Medchemexpress LLC BMS-202 | 1675203-84-5 | 99.4% | 419.52 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1974550MG
Encompass_Preferred
BMS-202 is a potent, nonpeptidic PD-1/PD-L1 complex inhibitor with an IC50 of 18 nM and a KD of 8 μM. It binds to PD-L1, blocking human PD-1/PD-L1 interaction, and exhibits antitumor activity.
  • Potent and nonpeptidic PD-1/PD-L1 complex inhibitor
  • Binds to PD-L1 and blocks human PD-1/PD-L1 interaction
  • Exhibits antitumor activity
  • Soluble in various in vitro and in vivo solutions
  • Stable under recommended storage conditions
  • Cited in numerous scientific publications
Catalog No. 50-004-01814