Learn More
Medchemexpress LLC BMS-209641 | 369364-50-1 | 98.1% | 50 MG

Supplier: Medchemexpress LLC HY11951850MG
BMS641 (BMS-209641) is a selective retinoic acid receptor β (RARβ) agonist used in biochemical and pharmacological research. It shows strong selectivity for RARβ over RARα and RARγ, and is provided as a white to off-white solid for use in in vitro and in vivo studies. Not for human use.
- Selective RARβ agonist with high affinity (Kd ≈ 2.5 nM)
- Approximately 100-fold selectivity versus RARα and RARγ
- High purity suitable for research applications
- Molecular weight 414.92 g/mol and formula C27H23ClO2
- Soluble in DMSO and formulatable for in vivo dosing
- Stable as a powder under recommended storage conditions
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.