Learn More
Medchemexpress LLC BMS-214662 (hydrochloride) | 195981-08-9 | 526.073 g/mol | 50 MG
SDP
Supplier:  Medchemexpress LLC HY16111A50MG
Encompass_Preferred
BMS-214662 hydrochloride is an inhibitor of farnesyltransferase. It can effectively block the localization and function of Ras proteins on the cell membrane by inhibiting the prenylation modification of Ras proteins, thereby exerting anti-tumor activity. It can be used in the research of tumor diseases related to Ras.
  • Inhibits farnesyltransferase
  • Blocks localization and function of Ras proteins
  • Exhibits anti-tumor activity
  • IC50 value for H-Ras is 1.3 nM
  • IC50 value for K-Ras is 8.4 nM
  • Suitable for research of tumor diseases related to Ras
Catalog No. 50-003-95099