Learn More
Medchemexpress LLC BMS-232632 sulfate | 229975-97-7 | 99.1% | 802.93 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY17367A100MG
Encompass_Preferred
Atazanavir sulfate is a highly selective and orally active HIV-1 protease inhibitor with blood-brain barrier permeability. It functions as both a substrate and inhibitor of CYP3A4 and an inhibitor of P-glycoprotein (P-gp). This compound also acts as a SARS-CoV 3CLpro inhibitor with an IC50 of 3.49 μM and has demonstrated efficacy in inhibiting cardiac fibrosis, hyperlipidemia, and inducing malignant glioma cell death.
  • Targets HIV-1 protease, CYP3A4, P-glycoprotein, and SARS-CoV 3CLpro.
  • Shows an IC50 of 3.49 μM for SARS-CoV 3CLpro.
  • Attenuates hypoxia-induced cardiac fibroblast proliferation.
  • Inhibits collagen I and III expression in cardiac fibroblasts.
  • Induces endoplasmic reticulum stress response in glioma cells.
  • Reduces collagen deposition and cardiac remodeling in MI models.
Catalog No. 50-003-84760