Learn More
Medchemexpress LLC BMS 310705 | 280578-49-6 | 99.1% | 522.70 g/mol | C27H42N2O6S | 10 MG

Supplier: Medchemexpress LLC HY10702010MG
BMS 310705 (21-Aminoepothilone B) is an epothilone-derived small-molecule apoptosis inducer used in biochemical and cellular research. It is characterized by CAS 280578-49-6, molecular formula C27H42N2O6S, and high reported purity, and is supplied as a solid optimized for DMSO solubility and long-term storage.
- Epothilone-derived apoptosis inducer for cell-based assays.
- High purity (99.1%) suitable for research applications.
- Molecular weight 522.70 g/mol for accurate dosing.
- Soluble in DMSO at 100 mg/mL with sonication.
- Powder stable at -20°C for up to 3 years.
- In vivo formulation achievable at ≥2.5 mg/mL in mixed solvent protocol.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.