Learn More
Medchemexpress LLC BMS-37 | 1675202-20-6 | C27H32N2O4 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY150879
Encompass_Preferred
BMS-37 is a PD-1/PD-L1 immune checkpoint inhibitor with an IC50 towards the PD-L1/PD-1 complex ranging from 18 to 200 nM. It exhibits unspecific toxicity against modified Jurkat T cells with an EC50 between 3 and 6 μM. It can be utilized for studying PD-L1-induced exhaustion of T-cells or as a PD-L1 ligand for synthesizing PROTAC molecules.
  • PD-1/PD-L1 immune checkpoint inhibitor.
  • Can be used for the study of PD-L1-induced exhaustion of T-cells.
  • Can be used as a PD-L1 ligand to synthesize PROTAC molecules.
Catalog No. 50-003-74334