Learn More
Cayman Chemical BMS 470539hydrochlorIde 1mg
SDP
Supplier:  Cayman Chemical 222311 MG
Encompass_Preferred
An MC1R agonist; induces cAMP accumulation in B16/F10 murine melanoma cells and in CHO cells overexpressing human MC1R (EC50s = 11.6 and 16.8 nM, respectively); inhibits LPS-induced TNF-α production in mice (ED50 = 10 μmol/kg, s.c.); reduces LPS-induced leukocyte infiltration and paw swelling in mouse models of lung inflammation and delayed-type hypersensitivity, respectively; decreases retinal damage in a mouse model of streptozotocin-induced diabetic retinopathy; increases striatal dopamine levels and decreases the number of striatal α-synuclein aggregates in a mouse model of Parkinson’s disease at 20 mg/kg
Catalog No. 50-296-4146