Learn More
Medchemexpress LLC Bms-502 | 2407854-18-4 | 99.3% | 516.50 | C27H22F2N6O3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY14987410MG
Encompass_Preferred
BMS-502 is a small-molecule research compound that acts as a potent dual inhibitor of diacylglycerol kinase (DGK) α and ζ, intended for preclinical and in vitro studies. It is provided with analytical data and storage recommendations to support research use.
  • Potent dual inhibition of DGK α and ζ (IC50 4.6 nM and 2.1 nM).
  • High purity suitable for research use (≈99.3%).
  • Molecular weight 516.50; chemical formula C27H22F2N6O3 for compound identification.
  • Available in small mass pack sizes convenient for screening and assay work.
  • Stable under recommended storage conditions (-20°C; solvent storage guidance provided).
Catalog No. 50-002-85771