Learn More
Medchemexpress LLC HY-11092 50mg Medchemexpress, BMS-509744 CAS:439575-02-7 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY1109250MG
Encompass_Preferred
Medchemexpress,HY-11092 50mg BMS-509744 CAS:439575-02-7 Formula:C32H41N5O4S2 BMS-509744 is a potent and selective Itk kinase inhibitor with an IC50 value of 19 nM. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-7482