Learn More
Medchemexpress LLC BMS-509744 | 439575-02-7 | 98.1% | 623.83 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY11092100MG
Encompass_Preferred
BMS-509744 is a potent, selective, and ATP-competitive Itk inhibitor with an IC50 of 19 nM. This compound reduces T-cell receptor-induced functions, including PLCγ1 tyrosine phosphorylation, calcium mobilization, IL-2 secretion, and T-cell proliferation in vitro in both human and mouse cells. It has also been shown to significantly diminish lung inflammation in a mouse model of ovalbumin-induced allergy/asthma.
  • Potent Itk inhibitor
  • Selective and ATP competitive
  • Reduces T-cell receptor-induced functions (e.g., PLCγ1 tyrosine phosphorylation, calcium mobilization, IL-2 secretion, T-cell proliferation)
  • Diminishes lung inflammation in mouse models of allergy/asthma
Catalog No. 50-003-43902