Learn More
Medchemexpress LLC BMS-582949 hydrochloride | 912806-16-7 | 98.9% | C22H27ClN6O2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY14305A100MG
Encompass_Preferred
BMS-582949 hydrochloride is an orally active and highly selective p38α MAPK inhibitor with an IC50 of 13 nM. It demonstrates a significantly improved pharmacokinetic profile and is effective in inflammatory diseases.
  • Target: p38α MAPK (IC50: 13 nM), TNFα (IC50: 50 nM, in cells)
  • Selectivity: exhibits >2000-fold selectivity for p38α over 57 diverse kinases
  • In vivo efficacy: effective in both acute murine inflammation models and rat adjuvant arthritis models
  • Orally active
  • High selectivity for p38α MAPK
  • Improved pharmacokinetic profile
  • Effective in inflammatory diseases
Catalog No. 50-003-53837