Learn More
Medchemexpress LLC BMS-582949 hydrochloride | 912806-16-7 | 98.90% | C22H27ClN6O2 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY14305A
Encompass_Preferred
BMS-582949 hydrochloride is an orally active and highly selective p38α MAPK inhibitor with an IC50 of 13 nM. It displays a significantly improved pharmacokinetic profile and is effective in inflammatory disease. BMS-582949 is a weak inhibitor of CYP3A4 and displays >2000-fold selectivity for p38α over a diverse panel of 57 kinases. In vivo, it is effective in both the acute murine model of inflammation and rat adjuvant arthritis model. Storage conditions: 4°C, sealed storage, away from moisture. In solvent: -80°C for 6 months; -20°C for 1 month (sealed storage, away from moisture).
  • Orally active and highly selective p38α MAPK inhibitor
  • Improved pharmacokinetic profile
  • Effective in inflammatory disease
  • Weak inhibitor of CYP3A4
  • Displays >2000-fold selectivity for p38α over 57 kinases
  • Effective in acute murine models of inflammation and rat adjuvant arthritis models
Catalog No. 50-003-54694