Learn More
Medchemexpress LLC BMS-593214 1mg | 1004551-40-9 | 505.56 | C31H27N3O4 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1145111MG
Encompass_Preferred
This compound is an active site-directed Factor VIIa (FVIIa) inhibitor with antithrombotic and antihaemostatic activity for preclinical research use. It inhibits human FVIIa and modulates the VIIa-activated substrate FX; in vivo studies show prevention of carotid artery and vena cava thrombosis. Supplied as a solid and as DMSO solutions; consult the datasheet for storage and handling.
  • High purity suitable for research applications.
  • Direct competitive inhibitor of human FVIIa.
  • Demonstrated antithrombotic activity in animal models.
  • Soluble in DMSO at high concentration; ultrasonic treatment may be required.
  • Available as solid quantities and as ready-to-use DMSO solutions.
  • Store powder and solutions at recommended temperatures to preserve stability.
Catalog No. 50-002-29139