Learn More
Medchemexpress LLC BMS-599626 (morpholin-3-ylmethyl carbamate) | 714971-09-2 | MFCD10565678 | 98.6% | 530.6 g/mol | C27H27FN8O3 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1025150MG
Encompass_Preferred
BMS-599626 (AC480) is a pan-HER kinase inhibitor used in research to target EGFR (HER1) and HER2 signaling. It is supplied as a white to off-white solid with high reported purity and recommended cold storage for stability.
  • Inhibits HER1 and HER2 with IC50s ≈ 20 nM and 30 nM respectively.
  • White to off-white solid with high purity (>98%).
  • Suitable for biochemical, cellular, and in vivo research applications.
  • Recommended storage: powder at -20°C for long term, 4°C for shorter storage.
  • Provided in milligram-scale packaging for research use.
Catalog No. 50-002-49004