Learn More
Selleck Chemical LLC BMS-687453 S0170-25MG
SDP
Supplier:  Selleck Chemical LLC S017025MG
Encompass_Preferred
BMS-687453 (compound 2) is a potent and selective agonist of PPAR with EC50 of 10 nM and IC50 of 260 nM for human PPAR
Catalog No. 50-000-40093