Learn More
Selleck Chemical LLC BMS-754807 S1124-50mg
SDP
Supplier:  Selleck Chemical LLC S112450MG
Encompass_Preferred
BMS-754807 is a potent and reversible inhibitor of IGF-1R/InsR with IC50 of 1 8 nM/1 7 nM in cell-free assays less potent to Met (c-Met) Aurora A/B TrkA/B and Ron and shows little activity to Flt3 Lck MK2 PKA PKC etc Phase 2
Catalog No. 50-310-8487