Learn More
Apexbio Technology LLC BMS-777607, 100mg. Cas: 1025720-94-8 MFCD: MFCD16495773. C-Met inhibitor, potent and selective
SDP
Supplier:  Apexbio Technology LLC A5703100
Encompass_Preferred
BMS 777607 is a novel, selective and orally available ATP-competitive MET kinase inhibitor that primarily targets several MET family members, including RON, MET, Tyro-3 and AxI. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4960