Learn More
Selleck Chemical LLC Daclatasvir (BMS-790052) 50mg 1009119-64-5 EBP883
SDP
Supplier:  Selleck Chemical LLC S148250MG
Encompass_Preferred
Daclatasvir (BMS-790052, EBP883) is a highly selective inhibitor of HCV NS5A with EC50 of 9-50 pM, for a broad range of HCV replicon genotypes and the JFH-1 genotype 2a infectious virus in cell culture. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-720-9