Learn More
Selleck Chemical LLC BMS-794833 50mg 1174046-72-0
SDP
Supplier:  Selleck Chemical LLC S220150MG
Encompass_Preferred
BMS-794833 is a potent ATP competitive inhibitor of Met (c-Met)/VEGFR2 with IC50 of 1.7 nM/15 nM, also inhibits Ron, Axl and Flt3 with IC50 of <3 nM; a prodrug of BMS-817378. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-737-0