Learn More
Medchemexpress LLC BMS-8 5mg | 1675201-90-7 | 494.42 g/mol | C27H28BrNO3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1162745MG
Encompass_Preferred
BMS-8 is a small-molecule inhibitor of the PD-1/PD-L1 protein-protein interaction. It binds directly to PD-L1 and induces PD-L1 homodimer formation, preventing PD-1 binding; reported IC50 is 7.2 μM. Supplied for research use in solid and solution formats.
Catalog No. 50-002-28971